2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Lieferant: FLUOROCHEM
Synonyme:
2-Brom-5-(trifluormethyl)benzol-1-sulfonylchlorid, 2-Brom-5-(trifluormethyl)benzolsulfochlorid, 2-Brom-5-(trifluormethyl)benzolsulfonsäurechlorid
013084-5G
013084-25G
FLUO013084-5GEA
249
EUR
FLUO013084-5G
FLUO013084-25G
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
2-Brom-5-(trifluormethyl)benzolsulfonylchlorid
Formel:
C₇H₃BrClF₃O₂S Siedepunkt: 230…231 °C (1013 hPa) |
MDL:
MFCD03094395 CAS-Nummer: 176225-08-4 UN: 3265 ADR: 8,II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...