4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 4-Nitro-3-(trifluoromethyl)benzene-1-sulphonyl chloride

FL13088.POR 013088-1G 013088-5G
CHMPFL13088.POREA 0 EUR
CHMPFL13088.POR FLUO013088-1G FLUO013088-5G
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
4-Nitro-3-(trifluormethyl)benzolsulfonylchlorid
Formel: C₇H₃ClF₃NO₄S
MDL: MFCD03094387
CAS-Nummer: 39234-83-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR