3-Brom-4-fluoranilin

Lieferant: FLUOROCHEM

013091-5G 013091-25G
FLUO013091-5GEA 45.9 EUR
FLUO013091-5G FLUO013091-25G
3-Brom-4-fluoranilin
Formel: C₆H₅BrFN
MDL: MFCD03788559
CAS-Nummer: 656-64-4
UN: 2811
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR