Dansylfluorid
Lieferant: FLUOROCHEM
Synonyme:
5-(Dimethylamino)naphthalin-1-sulfonylfluorid, 5-(Dimethylamino)naphthalinsulfonylfluorid, 5-(Dimethylamino)naphthalinsulfonsäurefluorid
013104-1G
FLUO013104-1GEA
479
EUR
FLUO013104-1G
Dansylfluorid
Dansylfluorid
Formel:
C₁₂H₁₂FNO₂S Molecular Weight: 253.3 g/mol |
MDL:
MFCD00042702 CAS-Nummer: 34523-28-9 EINECS: 252-071-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...