3-Cyan-4-fluoranilin

Lieferant: FLUOROCHEM

Synonyme: 5-Amino-2-fluorbenzonitril, 5-Amino-2-fluorbenzoesäurenitril

013105-5G 013105-25G 013105-100G
FLUO013105-5GEA 31.1 EUR
FLUO013105-5G FLUO013105-25G FLUO013105-100G
3-Cyan-4-fluoranilin
3-Cyan-4-fluoranilin
Formel: C₇H₅FN₂
MDL: MFCD00277872
CAS-Nummer: 53312-81-5
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR