2-Brom-3,5-difluorpyridin

Lieferant: FLUOROCHEM

013124-1G
FLUO013124-1GEA 67.1 EUR
FLUO013124-1G
2-Brom-3,5-difluorpyridin
2-Bromo-3,5-difluoropyridine
Formel: C₅H₂BrF₂N
MDL: MFCD03840197
CAS-Nummer: 660425-16-1

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR