2-Chlor-3-fluorisonicotinsäure

Lieferant: FLUOROCHEM

Synonyme: 2-Chloro-3-fluoropyridine-4-carboxylic acid

013130-1G
FLUO013130-1GEA 31.1 EUR
FLUO013130-1G
2-Chlor-3-fluorisonicotinsäure
2-Chlor-3-fluorisonicotinsäure
Formel: C₆H₃ClFNO₂
Molecular Weight: 175,55 g/mol
MDL: MFCD04038325
CAS-Nummer: 628691-93-0

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR