5-Iodisatin

Lieferant: FLUOROCHEM

013160-5G 013160-25G
FLUO013160-5GEA 82.1 EUR
FLUO013160-5G FLUO013160-25G
5-Iodisatin
5-Iodisatin
Formel: C₈H₄INO₂
MDL: MFCD00016899
CAS-Nummer: 20780-76-1
EINECS: 244-035-1

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR