1-(Trifluormethyl)cyclopropancarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 1-(Trifluoromethyl)cyclopropane-1-carboxylic acid

013181-1G 013181-2.5G 013181-5G 013181-100G
FLUO013181-1GEA 57.9 EUR
FLUO013181-1G FLUO013181-2.5G FLUO013181-5G FLUO013181-100G
1-(Trifluormethyl)cyclopropancarbonsäure
1-(Trifluoromethyl)cyclopropanecarboxylic acid
Formel: C₅H₅F₃O₂
Molecular Weight: 154.09 g/mol
MDL: MFCD03093070
CAS-Nummer: 277756-46-4
UN: 3261
ADR: 8 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR