2,3,6-Trifluor-4-(trifluormethyl)pyridin

Lieferant: FLUOROCHEM

013183-1G
FLUO013183-1GEA 0 EUR
FLUO013183-1G
2,3,6-Trifluor-4-(trifluormethyl)pyridin
2,3,6-Trifluoro-4-(trifluoromethyl)pyridine
Formel: C₆HF₆N
Molecular Weight: 201,07 g/mol
MDL: MFCD03840203
CAS-Nummer: 84940-46-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR