3-Brom-4-fluorbenzylamin Hydrochlorid

Lieferant: CHEMPUR

FL13223.POR 013223-25G 013223-5G
CHMPFL13223.POREA 0 EUR
CHMPFL13223.POR FLUO013223-25G FLUO013223-5G
3-Brom-4-fluorbenzylamin Hydrochlorid
3-Brom-4-fluorbenzylamin Hydrochlorid
Formel: C₇H₈BrClFN
Molecular Weight: 240.50 g/mol
MDL: MFCD00143094
CAS-Nummer: 202865-68-7

Bestellen

TECHNISCHE DATEN

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR