5-Fluor-2-methoxyphenol

Lieferant: CHEMPUR

Synonyme: 4-Fluor-2-hydroxyanisol, 1-Fluor-3-hydroxy-4-methoxybenzol

FL13351.POR 013351-25G 013351-5G
CHMPFL13351.POREA 0 EUR
CHMPFL13351.POR FLUO013351-25G FLUO013351-5G
5-Fluor-2-methoxyphenol
5-Fluor-2-methoxyphenol
Formel: C₇H₇FO₂
Molecular Weight: 142.13 g/mol
MDL: MFCD00143459
CAS-Nummer: 72955-97-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR