2-Brom-4,6-difluorbenzolsulfonylchlorid

Lieferant: CHEMPUR

Synonyme: 2-Brom-4,6-difluorbenzolsulfonsäurechlorid, 2-Brom-4,6-difluorbenzolsulfochlorid

FL13772.POR 013772-1G 013772-5G
CHMPFL13772.POREA 0 EUR
CHMPFL13772.POR FLUO013772-1G FLUO013772-5G
2-Brom-4,6-difluorbenzolsulfonylchlorid
2-Brom-4,6-difluorbenzolsulfonylchlorid
Formel: C₆H₂BrClF₂O₂S
Molecular Weight: 291.5 g/mol
Siedepunkt: 265…267 °C (1013 hPa)
Dichte: 1.93 g/cm³ (20 °C)
MDL: MFCD03094561
CAS-Nummer: 351003-42-4
UN: 3265
ADR: 8 II

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR