4-Brom-2,6-difluorbenzonitril

Lieferant: CHEMPUR

FL13773.POR 013773-1G 013773-5G
CHMPFL13773.POREA 0 EUR
CHMPFL13773.POR FLUO013773-1G FLUO013773-5G
4-Brom-2,6-difluorbenzonitril
4-Brom-2,6-difluorbenzonitril
Formel: C₇H₂BrF₂N
MDL: MFCD01310981
CAS-Nummer: 123843-67-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR