3-Fluor-4-hydroxybenzonitril

Lieferant: CHEMPUR

Synonyme: 3-Fluor-4-hydroxybenzoesäurenitril

FL13830.POR 013830-25G 013830-5G
CHMPFL13830.POREA 0 EUR
CHMPFL13830.POR FLUO013830-25G FLUO013830-5G
3-Fluor-4-hydroxybenzonitril
3-Fluor-4-hydroxybenzonitril
Formel: C₇H₄FNO
Molecular Weight: 137.11 g/mol
MDL: MFCD00215834
CAS-Nummer: 405-04-9
UN: 3439
ADR: 6.1 III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR