4-Fluor-3-nitrobenzonitril

Lieferant: CHEMPUR

FL13850.POR 013850-5G 013850-1G
CHMPFL13850.POREA 0 EUR
CHMPFL13850.POR FLUO013850-5G FLUO013850-1G
4-Fluor-3-nitrobenzonitril
4-Fluor-3-nitrobenzonitril
Formel: C₇H₃FN₂O₂
MDL: MFCD01632197
CAS-Nummer: 1009-35-4
UN: 3439
ADR: 6.1,III

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR