1-Iod-3-(trifluormethoxy)benzol

Lieferant: FLUOROCHEM

013870-1G 013870-5G 013870-25G
FLUO013870-1GEA 31.1 EUR
FLUO013870-1G FLUO013870-5G FLUO013870-25G
1-Iod-3-(trifluormethoxy)benzol
1-Iod-3-(trifluormethoxy)benzol
Formel: C₇H₄F₃IO
MDL: MFCD01090992
CAS-Nummer: 198206-33-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR