Ethyl-6-chlornicotinat
Lieferant: FLUOROCHEM
Synonyme:
Ethyl 6-chloronicotinate
013880-5G
013880-25G
FLUO013880-5GEA
0
EUR
FLUO013880-5G
FLUO013880-25G
Ethyl-6-chlornicotinat
Ethyl-6-chlornicotinat
Formel:
C₈H₈ClNO₂ |
MDL:
MFCD00082739 CAS-Nummer: 49608-01-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...