4,5,6-Trifluorpyrimidin
Lieferant: FLUOROCHEM
013885-1G
FLUO013885-1GEA
282
EUR
FLUO013885-1G
4,5,6-Trifluorpyrimidin
4,5,6-Trifluoropyrimidine
Formel:
C₄HF₃N₂ Molecular Weight: 134,06 g/mol |
MDL:
MFCD05662363 CAS-Nummer: 17573-78-3 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...