2-(Trifluormethyl)undecafluorpentan

Lieferant: FLUOROCHEM

Synonyme: Tetradecafluoro-2-methylpentane, Perfluoro-2-methylpentane, Perfluoroisohexane

013895-250G
FLUO013895-250GEA 189 EUR
FLUO013895-250G
2-(Trifluormethyl)undecafluorpentan
2-(Trifluormethyl)undecafluorpentan
Formel: C₆F₁₄
Molecular Weight: 338,04 g/mol
MDL: MFCD00059905
CAS-Nummer: 355-04-4
EINECS: 206-575-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR