3,4-Difluorphenylhydraziniumchlorid

Lieferant: FLUOROCHEM

Synonyme: 3,4-Difluorphenylhydrazin Hydrochlorid, 3,4-Difluorphenylhydrazinhydrochlorid, 1-(3,4-Difluorphenyl)hydrazin Hydrochlorid

013916-5G 013916-25G
FLUO013916-5GEA 36.2 EUR
FLUO013916-5G FLUO013916-25G
3,4-Difluorphenylhydraziniumchlorid
3,4-Difluorphenylhydraziniumchlorid
Formel: C₆H₇ClF₂N₂
Molecular Weight: 180.59 g/mol
MDL: MFCD03094170
CAS-Nummer: 875664-54-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR