Methyl-4-aminonicotinat

Lieferant: FLUOROCHEM

Synonyme: Methyl 4-aminonicotinate

013940-1G 013940-5G 013940-25G
FLUO013940-1GEA 31.1 EUR
FLUO013940-1G FLUO013940-5G FLUO013940-25G
Methyl-4-aminonicotinat
Methyl-4-aminonicotinat
Formel: C₇H₈N₂O₂
Molecular Weight: 152.15 g/mol
MDL: MFCD00955996
CAS-Nummer: 16135-36-7

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR