2-Brom-5-pyridinboronsäure
Lieferant: FLUOROCHEM
013947-1G
FLUO013947-1GEA
31.1
EUR
FLUO013947-1G
2-Brom-5-pyridinboronsäure
2-Brom-5-pyridinboronsäure
Formel:
C₅H₅BBrNO₂ |
MDL:
MFCD03411558 CAS-Nummer: 223463-14-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...