Methyl-2-amino-5-fluorbenzoat

Lieferant: FLUOROCHEM

013960-25G 013960-5G
FLUO013960-25GEA 146 EUR
FLUO013960-25G FLUO013960-5G
Methyl-2-amino-5-fluorbenzoat
Methyl-2-amino-5-fluorbenzoat
Formel: C₈H₈FNO₂
MDL: MFCD03791308
CAS-Nummer: 319-24-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR