2,4-Diethoxybenzaldehyd

Lieferant: FLUOROCHEM

014181-1G
FLUO014181-1GEA 0 EUR
FLUO014181-1G
2,4-Diethoxybenzaldehyd
2,4-Diethoxybenzaldehyde
Formel: C₁₁H₁₄O₃
Molecular Weight: 194,2271 g/mol
MDL: MFCD00995010
CAS-Nummer: 22924-16-9

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR