3,4-Diethoxybenzaldehyd
Lieferant: FLUOROCHEM
014187-1G
FLUO014187-1GEA
0
EUR
FLUO014187-1G
3,4-Diethoxybenzaldehyd
3,4-Diethoxybenzaldehyde
Formel:
C₁₁H₁₄O₃ Molecular Weight: 194,23 g/mol Siedepunkt: 279 °C (1013 hPa) Schmelzpunkt: 22 °C Dichte: 1,097 g/cm³ (25 °C) |
MDL:
MFCD00016607 CAS-Nummer: 2029-94-9 EINECS: 217-979-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...