4-(Benzyloxy)-3-methoxybenzaldehyd

Lieferant: FLUOROCHEM

014210-1G
FLUO014210-1GEA 0 EUR
FLUO014210-1G
4-(Benzyloxy)-3-methoxybenzaldehyd
4-(Benzyloxy)-3-methoxybenzaldehyd
Formel: C₁₅H₁₄O₃
MDL: MFCD00003365
CAS-Nummer: 2426-87-1
EINECS: 219-379-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR