2'-Methylbiphenyl-3-carbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2'-Methyl-[1,1'-biphenyl]-3-carboxylic acid
014238-1G
FLUO014238-1GEA
177
EUR
FLUO014238-1G
2'-Methylbiphenyl-3-carbonsäure
2'-Methylbiphenyl-3-carbonsäure
Formel:
C₁₄H₁₂O₂ Molecular Weight: 212,25 g/mol |
MDL:
MFCD03424589 CAS-Nummer: 168618-44-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...