2'-Methylbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Methyl-[1,1'-biphenyl]-4-carboxylic acid

014240-1G
FLUO014240-1GEA 177 EUR
FLUO014240-1G
2'-Methylbiphenyl-4-carbonsäure
2'-Methylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₂O₂
Molecular Weight: 212,25 g/mol
CAS-Nummer: 5748-43-6

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR