4'-Chlorbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Chloro[1,1'-biphenyl]-4-carboxylic acid, 4'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

014241-1G
FLUO014241-1GEA 82.1 EUR
FLUO014241-1G
4'-Chlorbiphenyl-4-carbonsäure
4'-Chlorbiphenyl-4-carbonsäure
Formel: C₁₃H₉ClO₂
MDL: MFCD03424592
CAS-Nummer: 5748-41-4

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR