2'-Chlorbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-Chloro-[1,1'-biphenyl]-4-carboxylic acid

014243-1G
FLUO014243-1GEA 264 EUR
FLUO014243-1G
2'-Chlorbiphenyl-4-carbonsäure
2'-Chlorbiphenyl-4-carbonsäure
Formel: C₁₃H₉ClO₂
Molecular Weight: 232,67 g/mol
MDL: MFCD03424594
CAS-Nummer: 3808-93-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR