3'-Chlorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014244-1G
FLUO014244-1GEA 189 EUR
FLUO014244-1G
3'-Chlorbiphenyl-3-carbonsäure
3'-Chlorbiphenyl-3-carbonsäure
Formel: C₁₃H₉ClO₂
MDL: MFCD03424595
CAS-Nummer: 168619-06-5

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR