2'-Chlorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014245-1G
FLUO014245-1GEA 264 EUR
FLUO014245-1G
2'-Chlorbiphenyl-3-carbonsäure
2'-Chlorbiphenyl-3-carbonsäure
Formel: C₁₃H₉ClO₂
MDL: MFCD03424596
CAS-Nummer: 168619-03-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR