3'-Methoxybiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3'-Methoxy-[1,1'-biphenyl]-3-carboxylic acid

014247-100MG 014247-1G
FLUO014247-100MGEA 0 EUR
FLUO014247-100MG FLUO014247-1G
3'-Methoxybiphenyl-3-carbonsäure
3'-Methoxybiphenyl-3-carbonsäure
Formel: C₁₄H₁₂O₃
Molecular Weight: 228.25 g/mol
MDL: MFCD03424598
CAS-Nummer: 168618-45-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR