2'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 2'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

014256-1G
FLUO014256-1GEA 534 EUR
FLUO014256-1G
2'-(Trifluormethyl)biphenyl-4-carbonsäure
2'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD03424605
CAS-Nummer: 198205-79-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR