4'-(Trifluormethyl)biphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-(Trifluoromethyl)-[1,1'-biphenyl]-4-carboxylic acid

014257-1G
FLUO014257-1GEA 113 EUR
FLUO014257-1G
4'-(Trifluormethyl)biphenyl-4-carbonsäure
4'-(Trifluormethyl)biphenyl-4-carbonsäure
Formel: C₁₄H₉F₃O₂
Molecular Weight: 266,22 g/mol
MDL: MFCD03424606
CAS-Nummer: 195457-71-7

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR