2'-Fluorbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014263-100MG 014263-1G
FLUO014263-100MGEA 0 EUR
FLUO014263-100MG FLUO014263-1G
2'-Fluorbiphenyl-3-carbonsäure
2'-Fluorbiphenyl-3-carbonsäure
Formel: C₁₃H₉FO₂
MDL: MFCD03424610
CAS-Nummer: 103978-23-0

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR