3',4'-Dimethoxybiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 3',4'-Dimethoxy-[1,1'-biphenyl]-3-carboxylic acid

014265-1G
FLUO014265-1GEA 369 EUR
FLUO014265-1G
3',4'-Dimethoxybiphenyl-3-carbonsäure
3',4'-Dimethoxybiphenyl-3-carbonsäure
Formel: C₁₅H₁₄O₄
Molecular Weight: 258,27 g/mol
MDL: MFCD03424611
CAS-Nummer: 676348-31-5

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR