3',4'-Dimethoxybiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

014266-1G
FLUO014266-1GEA 441 EUR
FLUO014266-1G
3',4'-Dimethoxybiphenyl-4-carbonsäure
3',4'-Dimethoxybiphenyl-4-carbonsäure
Formel: C₁₅H₁₄O₄
CAS-Nummer: 122294-10-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR