4'-Formylbiphenyl-4-carbonsäure

Lieferant: FLUOROCHEM

Synonyme: 4'-Formyl-[1,1'-biphenyl]-4-carboxylic acid

014267-1G
FLUO014267-1GEA 317 EUR
FLUO014267-1G
4'-Formylbiphenyl-4-carbonsäure
4'-Formylbiphenyl-4-carbonsäure
Formel: C₁₄H₁₀O₃
Molecular Weight: 226,2274 g/mol
MDL: MFCD03424613
CAS-Nummer: 70916-98-2

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR