2'-Formylbiphenyl-3-carbonsäure

Lieferant: FLUOROCHEM

014270-1G
FLUO014270-1GEA 536 EUR
FLUO014270-1G
2'-Formylbiphenyl-3-carbonsäure
2'-Formylbiphenyl-3-carbonsäure
Formel: C₁₄H₁₀O₃
MDL: MFCD03424616
CAS-Nummer: 205871-52-9

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR