5-Methyl-2-furancarbonsäure

Lieferant: FLUOROCHEM

Synonyme: 5-Methylfuran-2-carboxylic acid

014513-1G
FLUO014513-1GEA 44.6 EUR
FLUO014513-1G
5-Methyl-2-furancarbonsäure
5-Methyl-2-furoic acid
Formel: C₆H₆O₃
Molecular Weight: 126,11 g/mol
MDL: MFCD00092359
CAS-Nummer: 1917-15-3

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR