5-Methyl-2-thenoesäure

Lieferant: FLUOROCHEM

Synonyme: 5-Methylthiophene-2-carboxylic acid

014514-1G
FLUO014514-1GEA 0 EUR
FLUO014514-1G
5-Methyl-2-thenoesäure
5-Methyl-2-thenoesäure
Formel: C₆H₆O₂S
MDL: MFCD00005439
CAS-Nummer: 1918-79-2
EINECS: 217-640-3

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR