2-Methyl-3-furancarbonsäure
Lieferant: FLUOROCHEM
Synonyme:
2-Methylfuran-3-carboxylic acid
014518-1G
FLUO014518-1GEA
0
EUR
FLUO014518-1G
2-Methyl-3-furancarbonsäure
2-Methyl-3-furoic acid
Formel:
C₆H₆O₃ Molecular Weight: 126,11 g/mol |
MDL:
MFCD00092314 CAS-Nummer: 6947-94-0 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...