3-Chlor-2-benzo[b]thiophencarbonsäure
Lieferant: FLUOROCHEM
CFF217.10
014581-10G
CHMPCFF217.10EA
851
EUR
CHMPCFF217.10
FLUO014581-10G
3-Chlor-2-benzo[b]thiophencarbonsäure
3-Chlor-2-benzo[b]thiophencarbonsäure
Formel:
C₉H₅ClO₂S |
MDL:
MFCD00014579 CAS-Nummer: 21211-22-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...