3-Ethoxybenzhydrazid

Lieferant: FLUOROCHEM

Synonyme: 3-Ethoxybenzoic hydrazide

014598-1G
FLUO014598-1GEA 0 EUR
FLUO014598-1G
3-Ethoxybenzhydrazid
3-Ethoxybenzhydrazid
Formel: C₉H₁₂N₂O₂
Molecular Weight: 180.21 g/mol
MDL: MFCD00017063
CAS-Nummer: 27830-16-6

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR