5-Chlor-2-thenoesäure
Lieferant: FLUOROCHEM
Synonyme:
5-Chlor-2-thiophencarbonsäure, 5-Chlorthiophen-2-carbonsäure
014607-1G
014607.5G
FLUO014607-1GEA
76.2
EUR
FLUO014607-1G
FLUO014607.5G
5-Chlor-2-thenoesäure
5-Chlor-2-thenoesäure
Formel:
C₅H₃ClO₂S Molecular Weight: 162,6 g/mol |
MDL:
MFCD00041426 CAS-Nummer: 24065-33-6 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...