2-Phenyl-4-chinolincarbonsäurehydrazid

Lieferant: FLUOROCHEM

014613-1G
FLUO014613-1GEA 248 EUR
FLUO014613-1G
2-Phenyl-4-chinolincarbonsäurehydrazid
2-Phenylquinoline-4-carboxylic acid hydrazide
Formel: C₁₆H₁₃N₃O
MDL: MFCD00160604
CAS-Nummer: 4779-54-8

Bestellen


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR