4-Brom-1,5-dimethyl-1H-pyrazol-3-carbonsäurehydrazid

Lieferant: FLUOROCHEM

014629-1G
FLUO014629-1GEA 644 EUR
FLUO014629-1G
4-Brom-1,5-dimethyl-1H-pyrazol-3-carbonsäurehydrazid
4-Bromo-1,5-dimethyl-1H-pyrazole-3-carboxylic acid hydrazide
Formel: C₆H₉BrN₄O
MDL: MFCD00662638
CAS-Nummer: 261965-56-4

Bestellen

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Mehr About VWR