4-Butoxybenzoesäure
Lieferant: FLUOROCHEM
014641-1G
FLUO014641-1GEA
0
EUR
FLUO014641-1G
4-Butoxybenzoesäure
4-Butoxybenzoesäure
Formel:
C₁₁H₁₄O₃ Siedepunkt: 319 °C (1013 hPa) Schmelzpunkt: 149 °C |
MDL:
MFCD00002546 CAS-Nummer: 1498-96-0 EINECS: 216-104-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...